Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromotoluene, 99%

Catalog No. p-7082064
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 95-46-5 | C7H7Br | 171.04 g/mol

2-Bromotoluene was used in the synthesis of (±)-isocomene. Suzuki coupling of 2-bromotoluene with phenylboronic acid using palladium nanoparticles supported on alumina-based oxides as catalyst has been reported. 2-Bromotoluene is used in manufacturing pharmaceuticals (especially for nonsteroidal antiinflammatory drugs; Meclofenamic acid, Mefenamic acid) and other organic compounds

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromotoluene was used in the synthesis of (±)-isocomene. Suzuki coupling of 2-bromotoluene with phenylboronic acid using palladium nanoparticles supported on alumina-based oxides as catalyst has been reported. 2-Bromotoluen
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-46-5
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.04
MDL Number MFCD00000068
InChI Key QSSXJPIWXQTSIX-UHFFFAOYSA-N
Synonym 2-bromotoluene, o-bromotoluene, 2-methylbromobenzene, o-tolyl bromide, benzene, 1-bromo-2-methyl, toluene, o-bromo, 2-bromo-1-methylbenzene, 2-tolyl bromide, 1-methyl-2-bromobenzene, o-methylphenyl bromide
PubChem CID 7236
IUPAC Name 1-bromo-2-methylbenzene
SMILES CC1=CC=CC=C1Br

Specifications

Melting Point -27°C
Density 1.42
Boiling Point 179°C to 181°C
Flash Point 78°C (172°F)
Refractive Index 1.556
Quantity 25 g
Beilstein 1904176
Merck Index 14,1439
Solubility Information Soluble in alcohol,ether,acetone,and ethanol. Insoluble in water.
Formula Weight 171.04
Percent Purity 99%
Chemical Name or Material 2-Bromotoluene
Show More Show Less
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227;
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 202-421-7
RTECSNumber XS7965500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.