Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanone, ACS, 99+%

Catalog No. AA39119K7
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L

Maximum level of impurities: Color (APHA) 15, Residue after evaporation 0.0025%, Titratable acid 0.0005meq/g, H2O 0.20% | CAS: 78-93-3 | C4H8O | 72.11 g/mol

2-Butanone is used as a solvent and for extractions in the pharmaceutical industry. 2-butanone acts as oxidant for the iridium-catalyzed oxidations of primary alcohols. It is useful for the catalytic transfer hydrogenation on MgO.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Synonym 2-butanone, methyl ethyl ketone, butanone, ethyl methyl ketone, methylethyl ketone, meetco, methyl acetone, methylethylketone, 3-butanone, ethylmethylketon
PubChem CID 6569
ChEBI CHEBI:28398
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

Melting Point °C to 87°C
Color Colorless
Appearance Colorless liquid
Density 0.805 g/cm3 at 20°C
Boiling Point 80°C
Quantity 4 L
Flash Point -3°C (26°F)
Ignition Point 514°C (957°F)
Odor Characteristic
Physical Form Liquid
Refractive Index 1.379
Assay Percent Range >99%
Vapor Pressure 101 hPa (76mm Hg) at 20°C
Beilstein 741880
Solubility Information Miscible in cold water,diethyl ether and acetone.
Formula Weight 72.11
Grade ACS
Chemical Name or Material 2-Butanone
Show More Show Less
Hazard Category H225-H319-H336-H373
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
DOTInformation DOT Class: 3, Packaging Group: II
EINECSNumber 201-159-0
RTECSNumber EL6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.