Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Butanone, CHROMASOLV™, for HPLC, ≥99.7%, Honeywell Riedel-de Haen
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Melting Point | -87°C (literature) |
| CAS Max % | 100.0000% |
| Density | 0.805 g/mL at 25 °C(lit.) |
| Boiling Point | 79°C to 81°C |
| Quantity | 4 x 2.5 L |
| Flash Point | −4°C |
| Packaging | Glass Bottle |
| Linear Formula | C2H5COCH3 |
| UN Number | UN1193 |
| Beilstein | 741880 |
| Show More |
Safety and Handling
| ShelfLife | 1260 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Ethyl methyl ketone |
| EINECSNumber | 201-159-0 |
| Recommended Storage | Room Temp |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction