Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butanone, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18600102
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can

M1260, 78-93-3, C4H8O

Spectrum™ Chemical 2-Butanone, Reagent, ACS

Chemical Identifiers

CAS 78-93-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00011648
InChI Key ZWEHNKRNPOVVGH-UHFFFAOYSA-N
IUPAC Name butan-2-one
SMILES CCC(C)=O

Specifications

CAS Max % 100%
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 0.0025%
Titratable Acid 0.0005 meq/g
UN Number UN1193
Water 0.2%
Percent Purity 99%
Grade ACS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.