Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Carboxybenzeneboronic acid, 95%

Catalog No. AAL1630103
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1630103 1 g
AAL16301MD 250 mg
AAL1630106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1630103 Supplier Thermo Scientific Chemicals Supplier No. L1630103
Add to Cart

CAS: 149105-19-1 | C7H7BO4 | 165.939 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149105-19-1
Molecular Formula C7H7BO4
Molecular Weight (g/mol) 165.939
MDL Number MFCD01318118
InChI Key KWNPRVWFJOSGMZ-UHFFFAOYSA-N
Synonym 2-carboxyphenylboronic acid, 2-dihydroxyboranyl benzoic acid, 2-carboxybenzeneboronic acid, 2-dihydroxyboronyl benzoic acid, 2-dihydroxyboryl benzoic acid, carboxyphenylboronic acid, benzoic acid, 2-borono, o-carboxyphenylboronic acid, 2-carboxyphenylboronicacid, borobenzoesaure
PubChem CID 2733985
IUPAC Name 2-boronobenzoic acid
SMILES B(C1=CC=CC=C1C(=O)O)(O)O

Specifications

Melting Point ∼220°C
Quantity 1 g
Beilstein 2835407
Formula Weight 165.94
Percent Purity 95%
Chemical Name or Material 2-Carboxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.