Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Checkers 2-Channel Rubber Duct Office Cable Protector
Shop All Justrite Safety Group Products
Click to view available options
Length (English):
5 ft.
10 ft.
Description
- Two-Channel Protector Multiple Communication Lines Or 14 Through 22 Gauge Wire
- Can Be Slit If Necessary (Supplied Unslit)
- Reduces Tripping Hazards
- Protects And Organizes Portable Cords And Cables
- Non-Metallic, Industrial-Grade Rubber Construction
- Multiple Sizes For Various Cable Diameters
- Grooved Underside For Better Traction
- Optional Connector Kit Available For Most Sizes
Specifications
Specifications
| Color | Black |
| Material | Rubber |
| Product Type | Cable Protector |
| Length (English) | 5 ft. |
| Length (Metric) | 1.5 m |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction