Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1-methylpyridinium iodide, 97%

Catalog No. AC189260250
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 14338-32-0 | C6H7ClIN | 255.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14338-32-0
Molecular Formula C6H7ClIN
Molecular Weight (g/mol) 255.48
MDL Number MFCD00011984
InChI Key ABFPKTQEQNICFT-UHFFFAOYSA-M
Synonym 2-chloro-1-methylpyridinium iodide, 2-chloro-1-methylpyridin-1-ium iodide, pyridinium, 2-chloro-1-methyl-, iodide, unii-rlq88nyz68, rlq88nyz68, 2-chloro-1-methylpyridiniumiodide, 2-chloro-1-methyl-pyridinium iodide, pubchem16659, acmc-1brmd, ksc494o1l
PubChem CID 167069
IUPAC Name 2-chloro-1-methylpyridin-1-ium;iodide
SMILES [I-].C[N+]1=CC=CC=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 195°C to 200°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96.0 to 104% (Argentometry)
Packaging Glass bottle
Quantity 25 g
Fieser 08,95; 12,116
Merck Index 15, 6387
Solubility Information Solubility in water: soluble
Formula Weight 255.49
Percent Purity 97%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2-Chloro-1-methylpyridinium iodide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 238-288-7
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.