Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1-phenothiazin-10-yl-propan-1-one, 95%, Thermo Scientific™

Catalog No. AC438330010
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 38076-63-0
Molecular Formula C15H12ClNOS
Molecular Weight (g/mol) 289.79
InChI Key NFDHJLPQTVVBOU-UHFFFAOYSA-N
Synonym 2-chloro-1-10h-phenothiazin-10-yl propan-1-one, 2-chloro-1-phenothiazin-10-yl-propan-1-one, 2-chloro-1-phenothiazin-10-yl propan-1-one, 10h-phenothiazine, 10-2-chloro-1-oxopropyl, 10-2-chloropropanoyl-10h-phenothiazine, 1-propanone,2-chloro-1-10h-phenothiazin-10-yl
PubChem CID 352185
IUPAC Name 2-chloro-1-phenothiazin-10-ylpropan-1-one
SMILES CC(C(=O)N1C2=CC=CC=C2SC3=CC=CC=C31)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 117.4°C to 120.4°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 289.79
Percent Purity 95%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-chloro-1-phenothiazin-10-yl-propan-1-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.