Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-2-ethoxyacetic acid ethyl ester, 90%, remainder mainly ethyl diethoxyacetat, Thermo Scientific Chemicals

Catalog No. AC279871000
Change view
Click to view available options
Quantity:
50 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC279871000 100 g
AC279870500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC279871000 Supplier Thermo Scientific Chemicals Supplier No. 279871000

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34006-60-5
Molecular Formula C6H11ClO3
Molecular Weight (g/mol) 166.60
MDL Number MFCD00043953
InChI Key WDCHMODANTVRTF-UHFFFAOYNA-N
Synonym 2-chloro-2-ethoxyacetic acid ethyl ester, ethyl 2-chloro-2-ethoxy-acetate, ethyl2-chloro-2-ethoxyacetate, ethyl chloro ethoxy acetate, chloro ethoxy acetic acid ethyl ester, ethyl alpha-chloro-alpha-ethoxyacetate, ethyl alpha-chloro-alpha-ethoxy-acetate, 2-chloro-2-ethoxy-acetic acid ethyl ester, acetic acid,2-chloro-2-ethoxy-, ethyl ester
PubChem CID 2724873
IUPAC Name ethyl 2-(1-chloroethoxy)acetate
SMILES CCOC(=O)COC(C)Cl

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Brown-Orange to Colorless
Infrared Spectrum Authentic
Assay Percent Range 90%
Linear Formula C2H5O(Cl)CHC(O)OC2H5
Quantity 100 g
Formula Weight 166.6
Percent Purity ≥88%
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-ethoxyacetic acid ethyl ester
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.