Learn More
2-Chloro-2-methylbutane, 95%
Description
2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.
Solubility
Soluble in ethanol, ethyl ether. Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents. Avoid extremes of temperature and direct sunlight.

Specifications
Specifications
Melting Point | -73°C |
Density | 0.866 |
Boiling Point | 85°C to 86°C |
Flash Point | −10°C (14°F) |
Refractive Index | 1.405 |
Quantity | 100 g |
UN Number | UN1107 |
Beilstein | 1696930 |
Solubility Information | Soluble in ethanol,ethyl ether. Insoluble in water. |
Formula Weight | 106.6 |
Show More |
Safety and Handling
Hazard Category | H225 |
Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapor. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL CHLORIDES |
EINECSNumber | 209-836-2 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.