Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-2-methylnonane, 90+%

Catalog No. AA4378414
Click to view available options
Quantity:
5 g
25 g

CAS: 4325-50-2 | C10H21Cl | 176.728 g/mol

It is used in the photoelectron spectroscopy of C60Fn- and C60Fm2 in the gas phase and the generation and characterization of C1-C60F47- and D2-C60F44 in solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4325-50-2
Molecular Formula C10H21Cl
Molecular Weight (g/mol) 176.728
MDL Number MFCD00060781
InChI Key QDJXQCKHBZEIJM-UHFFFAOYSA-N
Synonym dimethyloctyl chloride, acmc-20ao0l, 2-chloro-2-methyl-nonane, nonane,2-chloro-2-methyl
PubChem CID 138189
IUPAC Name 2-chloro-2-methylnonane
SMILES CCCCCCCC(C)(C)Cl

Specifications

Color Colorless
Quantity 25 g
UN Number UN3082
Formula Weight 176.73
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-methylnonane
Hazard Category H351
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.