Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3,4-dimethoxybenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. C199725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C199725G 25 g
C19975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C199725G Supplier TCI America Supplier No. C199725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5417-17-4
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
InChI Key SAWHDJTZESXNMM-UHFFFAOYSA-N
PubChem CID 79445
IUPAC Name 2-chloro-3,4-dimethoxybenzaldehyde
SMILES COC1=C(C(=C(C=C1)C=O)Cl)OC

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 25 g
Synonym 2-chloroveratraldehyde, 2-chloro-3,4-dimethoxy-benzaldehyde, benzaldehyde, 2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxy benzaldehyde, chloroveratraldehyde, pubchem7759, acmc-1aumn, 2-chloro-3,4 dimethoxybenzaldehyde, benzaldehyde,2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxybenzaldehyde
Formula Weight 200.62
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3,4-dimethoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.