Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-ethylbenzoxazolium Tetrafluoroborate 97.0+%, TCI America™
SDP

Catalog No. C090525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C090525G 25 g
C09055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C090525G Supplier TCI America Supplier No. C090525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 63212-53-3
Molecular Formula C9H9BClF4NO
Molecular Weight (g/mol) 269.431
MDL Number MFCD00011943
InChI Key MLQZANYFUYLJRK-UHFFFAOYSA-N
Synonym 2-chloro-3-ethylbenzoxazolium tetrafluoroborate, 2-chloro-3-ethylbenzo d oxazol-3-ium tetrafluoroborate, 2-chloro-3-ethyl-1,3-benzoxazol-3-ium tetrafluoroborate, 2-chloro-3-ethyl-benzooxazole; tetrafluoroboron, acmc-209ndy, 3-ethyl-2-chlorobenzoxazolium tetrafluoroborate, 2-chloro-3-ethylbenzoxazo-lium tetrafluoroborate
PubChem CID 2724240
IUPAC Name 2-chloro-3-ethyl-1,3-benzoxazol-3-ium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CC[N+]1=C(OC2=CC=CC=C21)Cl

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 25 g
UN Number 1759
Formula Weight 269.43
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-ethylbenzoxazolium Tetrafluoroborate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.