Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-fluoro-5-methylpyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAH6197203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6197203 1 g
AAH6197206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6197203 Supplier Thermo Scientific Chemicals Supplier No. H6197203

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34552-15-3
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD06658238
InChI Key XZJURWKNRAGMCG-UHFFFAOYSA-N
Synonym 2-chloro-3-fluoro-5-picoline, 2-chloro-3-fluoro-5-methyl-pyridine, 6-chloro-5-fluoro-3-picoline, pyridine, 2-chloro-3-fluoro-5-methyl, 2-chloro-3-fluoro-5-methyl pyridine, pubchem14231, ksc495q2t, 6-chloro-5-fluoro-3-methylpyridine
PubChem CID 13541365
IUPAC Name 2-chloro-3-fluoro-5-methylpyridine
SMILES CC1=CC(F)=C(Cl)N=C1

Specifications

Color White
Quantity 1 g
Formula Weight 145.56
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Chloro-3-fluoro-5-methylpyridine
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.