Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-pyridinol, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
6636-78-8
C5H4ClNO
129.55
MFCD00006235
RSOPTYAZDFSMTN-UHFFFAOYSA-N
2-chloro-3-hydroxypyridine,2-chloro-3-pyridinol,3-pyridinol, 2-chloro,unii-vy212q0s3j,2-chloro-3-hydroxy pyridine,2-chloro-3-hydroxypyridine 2-chloro-3-pyridinol,2-chlorpyridin-3-ol,pubchem2577,2-chloro-pyridin-3-ol,2-chloro-pyridine-3-ol
81136
2-chloropyridin-3-ol
C1=CC(=C(N=C1)Cl)O
6636-78-8
C5H4ClNO
129.55
MFCD00006235
RSOPTYAZDFSMTN-UHFFFAOYSA-N
2-chloro-3-hydroxypyridine,2-chloro-3-pyridinol,3-pyridinol, 2-chloro,unii-vy212q0s3j,2-chloro-3-hydroxy pyridine,2-chloro-3-hydroxypyridine 2-chloro-3-pyridinol,2-chlorpyridin-3-ol,pubchem2577,2-chloro-pyridin-3-ol,2-chloro-pyridine-3-ol
81136
2-chloropyridin-3-ol
C1=CC(=C(N=C1)Cl)O
Color Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 168°C to 172°C
Quantity 25g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula Cl(C5H3N)OH
Beilstein 21, II, 34
Formula Weight 129.55
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-chloro-3-pyridinol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.