Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4'-fluoroacetophenone, 99%

Catalog No. p-7026272
Click to view available options
Quantity:
25 g
100 g

CAS: 456-04-2 | C8H6ClFO | 172.583 g/mol

1-methyl-2-(4-methylthio)phenyl-4-(4-fluoro)phenylimidazole which was synthesized by condensing N-methyl-4-(methylthio)benzamidine with 2-chloro-4?-fluoroacetophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-methyl-2-(4-methylthio)phenyl-4-(4-fluoro)phenylimidazole which was synthesized by condensing N-methyl-4-(methylthio)benzamidine with 2-chloro-4′-fluoroacetophenone.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-04-2
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.583
MDL Number MFCD00011652
InChI Key UJZWJOQRSMOFMA-UHFFFAOYSA-N
Synonym 2-chloro-4'-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethanone, p-fluorophenacyl chloride, 4-fluorophenacyl chloride, alpha-chloro-4-fluoroacetophenone, 4-chloroacetyl-1-fluorobenzene, alpha-chloro-p-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethan-1-one, ethanone, 2-chloro-1-4-fluorophenyl, 2'-chloro-4-fluoroacetophenone
PubChem CID 120248
IUPAC Name 2-chloro-1-(4-fluorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CCl)F

Specifications

Melting Point 46°C to 50°C
Boiling Point 247°C
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN3261
Beilstein 637860
Solubility Information Insoluble in water.
Formula Weight 172.59
Percent Purity 99%
Chemical Name or Material 2-Chloro-4'-fluoroacetophenone
Hazard Category H301-H314-H317-H330
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 207-256-4
RTECSNumber AM6550000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.