Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4'-fluoroacetophenone 98.0+%, TCI America™
SDP

Catalog No. C197725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C197725G 25 g
C19775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C197725G Supplier TCI America Supplier No. C197725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 456-04-2
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.583
InChI Key UJZWJOQRSMOFMA-UHFFFAOYSA-N
Synonym 2-chloro-4'-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethanone, p-fluorophenacyl chloride, 4-fluorophenacyl chloride, alpha-chloro-4-fluoroacetophenone, 4-chloroacetyl-1-fluorobenzene, alpha-chloro-p-fluoroacetophenone, 2-chloro-1-4-fluorophenyl ethan-1-one, ethanone, 2-chloro-1-4-fluorophenyl, 2'-chloro-4-fluoroacetophenone
PubChem CID 120248
IUPAC Name 2-chloro-1-(4-fluorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CCl)F

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 247°C
Quantity 25 g
Formula Weight 172.58
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4′-fluoroacetophenone
RTECSNumber AM6550000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.