Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-fluorobenzoic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2252-51-9
C7H4ClFO2
174.56
MFCD00010615
GRPWQLDSGNZEQE-UHFFFAOYSA-N
benzoic acid, 2-chloro-4-fluoro,2-chloro-4-fluoro-benzoic acid,2-chloro-4-fluor obenzoic acid,pubchem1370,acmc-209fx5,ksc207e4n,rarechem al bo 0405,2-chloro 4-fluoro benzoic acid,2-chloro-4-fluoro benzoic acid,4-fluoro-2-chloro-benzoic acid
75259
2-chloro-4-fluorobenzoic acid
C1=CC(=C(C=C1F)Cl)C(=O)O
2252-51-9
C7H4ClFO2
174.56
MFCD00010615
GRPWQLDSGNZEQE-UHFFFAOYSA-N
benzoic acid, 2-chloro-4-fluoro,2-chloro-4-fluoro-benzoic acid,2-chloro-4-fluor obenzoic acid,pubchem1370,acmc-209fx5,ksc207e4n,rarechem al bo 0405,2-chloro 4-fluoro benzoic acid,2-chloro-4-fluoro benzoic acid,4-fluoro-2-chloro-benzoic acid
75259
2-chloro-4-fluorobenzoic acid
C1=CC(=C(C=C1F)Cl)C(=O)O
Melting Point 183°C to 186°C
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula ClC6H3(F)CO2H
Formula Weight 174.56
Physical Form Powder or Flakes
Percent Purity 99%
Chemical Name or Material 2-Chloro-4-fluorobenzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-845-0

TSCA : TSCA