Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-fluorobenzoic acid, 99%, Thermo Scientific™

Catalog No. p-87271
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 2252-51-9
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.56
MDL Number MFCD00010615
InChI Key GRPWQLDSGNZEQE-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-4-fluoro, 2-chloro-4-fluoro-benzoic acid, 2-chloro-4-fluor obenzoic acid, pubchem1370, acmc-209fx5, ksc207e4n, rarechem al bo 0405, 2-chloro 4-fluoro benzoic acid, 2-chloro-4-fluoro benzoic acid, 4-fluoro-2-chloro-benzoic acid
PubChem CID 75259
IUPAC Name 2-chloro-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)Cl)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 183°C to 186°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(F)CO2H
Quantity 5 g
Formula Weight 174.56
Percent Purity 99%
Physical Form Powder or Flakes
Chemical Name or Material 2-Chloro-4-fluorobenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-845-0
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.