Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4'-fluorobenzophenone 98.0+%, TCI America™
SDP

Catalog No. C186025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C186025G 25 g
C18605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C186025G Supplier TCI America Supplier No. C186025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1806-23-1
Molecular Formula C13H8ClFO
Molecular Weight (g/mol) 234.654
MDL Number MFCD00000559
InChI Key DODIKYQYCCFWRZ-UHFFFAOYSA-N
PubChem CID 74547
IUPAC Name (2-chlorophenyl)-(4-fluorophenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)F)Cl

Specifications

Melting Point 62°C
Quantity 25 g
Formula Weight 234.65
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4′-fluorobenzophenone
EINECSNumber (4)-1488
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.