Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-iodo-3-pyridinemethanol, 97%, Thermo Scientific Chemicals

Catalog No. AAH5005203
Click to view available options
Quantity:
1 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 884494-44-4
Molecular Formula C6H5ClINO
Molecular Weight (g/mol) 269.466
MDL Number MFCD03095296
InChI Key BZQRQPAKTCMYIA-UHFFFAOYSA-N
Synonym 2-chloro-4-iodopyridin-3-yl methanol, 2-chloro-4-iodo-3-pyridinemethanol, 2-chloro-3-hydroxymethyl-4-iodopyridine, 2-chloro-4-iodo-pyridin-3-yl-methanol, acmc-20anzo, pubchem17089, 3-pyridinemethanol,2-chloro-4-iodo, 2-chloro-4-iodo-3-pyridyl methan-1-ol
PubChem CID 40418628
IUPAC Name (2-chloro-4-iodopyridin-3-yl)methanol
SMILES C1=CN=C(C(=C1I)CO)Cl

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 269.47
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-iodo-3-pyridinemethanol
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.