Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-nitropyridine N-Oxide 96.0+%, TCI America™
SDP

Catalog No. C265425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C265425G 25 g
C26545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C265425G Supplier TCI America Supplier No. C265425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14432-16-7
Molecular Formula C5H3ClN2O3
Molecular Weight (g/mol) 174.54
MDL Number MFCD00955617
InChI Key YSTCMHHKDOVZDA-UHFFFAOYSA-N
PubChem CID 84433
IUPAC Name 2-chloro-4-nitro-1-oxidopyridin-1-ium
SMILES C1=C[N+](=C(C=C1[N+](=O)[O-])Cl)[O-]

Specifications

Melting Point 150°C
Color Yellow
Quantity 25 g
UN Number 2811
Synonym 2-chloro-4-nitropyridine-n-oxide, 2-chloro-4-nitropyridine 1-oxide, 2-chloro-4-nitropyridine n-oxide, 2-chloro-4-nitropyridine1-oxide, 2-chloro-4-nitropyridin-1-ium-1-olate, 2-chloro-4-nitropyridin-1-ol, 2-chlor-4-nitropyridin-1-oxid, pubchem1191, acmc-1cisj, pycl12
Formula Weight 174.54
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-nitropyridine N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.