Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4'-phenylacetophenone 99.0+%, TCI America™
SDP

Catalog No. C025225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C025225G 25 g
C02525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C025225G Supplier TCI America Supplier No. C025225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 635-84-7
Molecular Formula C14H11ClO
Molecular Weight (g/mol) 230.69
MDL Number MFCD00058937
InChI Key IQEIGQFNDLINOT-UHFFFAOYSA-N
Synonym 4-Chloroacetylbiphenyl, 4-Phenylphenacyl Chloride
PubChem CID 69463
IUPAC Name 1-{[1,1'-biphenyl]-4-yl}-2-chloroethan-1-one
SMILES ClCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 126°C
Color White-Yellow
Quantity 25 g
Formula Weight 230.69
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4′-phenylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.