Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-fluoroaniline, 97%

Catalog No. AAB2561806
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 452-83-5 | C6H5ClFN | 145.561 g/mol

2-Chloro-5-fluoroaniline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-5-fluoroaniline is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-83-5
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.561
MDL Number MFCD00084852
InChI Key VWUFOZAFKYOZJB-UHFFFAOYSA-N
Synonym 5-fluoro-2-chloroaniline, 2-chloro-5-fluorobenzenamine, 2-chloro-5-fluoro-phenylamine, 2-chloranyl-5-fluoranyl-aniline, pubchem1517, acmc-1bn9k, 2-chloro-5-fluoro-aniline, 2-chloro-5-fluorophenylamine, ksc235o7b, 2-chloro-5-fluorobenzeneamine
PubChem CID 2736509
IUPAC Name 2-chloro-5-fluoroaniline
SMILES C1=CC(=C(C=C1F)N)Cl

Specifications

Melting Point ∼26°C
Boiling Point 211°C
Quantity 5 g
Odor Characteristic
Refractive Index 1.556
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 145.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-fluoroaniline
Hazard Category H302+H332-H311-H315-H319-H373
Hazard Statement GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.