Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-fluoropyridine, 95%, Thermo Scientific™

Catalog No. AC442860050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442860050 5 g
AC442860010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442860050 Supplier Thermo Scientific Chemicals Supplier No. 442860050

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Chemical Identifiers

CAS 31301-51-6
Molecular Formula C5H3ClFN
Molecular Weight (g/mol) 131.53
MDL Number MFCD03095332
InChI Key QOGXQLSFJCIDNY-UHFFFAOYSA-N
Synonym 2-chloro-5-fluoro-pyridine, 2-chloro-5-fluoro pyridine, pyridine, 2-chloro-5-fluoro, zlchem 338, pubchem1097, 2-chlor-5-fluorpyridin, acmc-1aevz, 2-chloro-5-fluropyridine, ksc223e3t, tpc-py082
PubChem CID 2782801
IUPAC Name 2-chloro-5-fluoropyridine
SMILES FC1=CN=C(Cl)C=C1

Specifications

Melting Point 23°C to 24°C
Density 1.2970g/mL
Boiling Point 74°C (37.0 Torr)
Flash Point 42°C
Assay Percent Range 94% min. (GC)
Quantity 5 g
Specific Gravity 1.297
Formula Weight 131.54
Percent Purity 95%
Chemical Name or Material 2-Chloro-5-fluoropyridine
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.