Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-iodo-3-(trifluoromethyl)pyridine, 95%

Catalog No. AAH6432906
Click to view available options
Quantity:
1 g
5 g

CAS: 887707-25-7 | C6H2ClF3IN | 307.44 g/mol

It is used as pharmaceutical intermediate and as an advanced chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate and as an advanced chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 887707-25-7
Molecular Formula C6H2ClF3IN
Molecular Weight (g/mol) 307.44
MDL Number MFCD12024302
InChI Key KUASPTUGCBGXAE-UHFFFAOYSA-N
Synonym 2-chloro-5-iodo-3-trifluoromethyl pyridine, 2-chloro-3-trifluoromethyl-5-iodo-pyridine, 2-chloro-3-trifluoromethyl-5-iodopyridine, 2-choro-5-iodo-3-trifluoromethylpyridine, 2-chloro-3-trifluoromethyl-5-iodo pyridine
PubChem CID 11500513
IUPAC Name 2-chloro-5-iodo-3-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC(I)=CN=C1Cl

Specifications

Melting Point 43°C
Quantity 5 g
Beilstein 10249610
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 307.44
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Chloro-5-iodo-3-(trifluoromethyl)pyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.