Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-iodobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. C219625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C219625G 25 g
C21965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C219625G Supplier TCI America Supplier No. C219625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 260355-20-2
Molecular Formula C7H3ClF3I
Molecular Weight (g/mol) 306.45
MDL Number MFCD09037801
InChI Key MCIFWVOVVPVBRJ-UHFFFAOYSA-N
Synonym 1-Chloro-4-iodo-2-(trifluoromethyl)benzene
PubChem CID 43145392
IUPAC Name 1-chloro-4-iodo-2-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=C(Cl)C=CC(I)=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 306.45
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2-Chloro-5-iodobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.