Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-methoxyaniline Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C155525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C155525G 25 g
C15555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C155525G Supplier TCI America Supplier No. C155525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85006-21-9
Molecular Formula C7H9Cl2NO
Molecular Weight (g/mol) 194.06
MDL Number MFCD00012962
InChI Key NQWBPXKJBZYGHZ-UHFFFAOYSA-N
Synonym 2-chloro-5-methoxyaniline hydrochloride, 3-amino-4-chloroanisole hydrochloride, 6-chloro-m-anisidine hydrochloride, 2-chloro-5-methoxyaniline hcl, 6-chloro-meta-anisidine hydrochloride, 2-chloro-5-methoxyanilinehydrochloride, benzenamine, 2-chloro-5-methoxy-, hydrochloride, timtec-bb sbb008038, pubchem3658, pubchem3660
PubChem CID 2723900
IUPAC Name 2-chloro-5-methoxyanilinium chloride
SMILES [Cl-].COC1=CC=C(Cl)C([NH3+])=C1

Specifications

Quantity 25 g
Formula Weight 194.06
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-methoxyaniline Hydrochloride
EINECSNumber (3)-0908
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.