Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-methylthiophene, 97%

Catalog No. AAB2165714
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAB2165714

Supplier: Thermo Scientific Chemicals B2165714

Only null left
Add to Cart

CAS: 17249-82-0 | C5H5ClS | 132.605 g/mol

It is used in the synthesis of dithienylcyclopentene optical molecular switches. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of dithienylcyclopentene optical molecular switches. It is also used as an active pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17249-82-0
Molecular Formula C5H5ClS
Molecular Weight (g/mol) 132.605
MDL Number MFCD01632145
InChI Key JSMMZMYGEVUURX-UHFFFAOYSA-N
Synonym thiophene, 2-chloro-5-methyl, pubchem5197, acmc-209e4p, 2-chloro-5-methyl-thiophene, ksc497i0h, 2-chloranyl-5-methyl-thiophene
PubChem CID 140208
IUPAC Name 2-chloro-5-methylthiophene
SMILES CC1=CC=C(S1)Cl

Specifications

Density 1.21
Boiling Point 155°C to 156°C
Flash Point 47°C (117°F)
Odor Characteristic
Refractive Index 1.5356
Quantity 25 g
UN Number UN1992
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 132.61
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-methylthiophene
Show More Show Less
Hazard Category H226-H302+H312-H331
Hazard Statement GHS H Statement
H331-H226-H302-H312
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.