Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-nitro-4-picoline, 98%, Thermo Scientific™

Catalog No. p-89532
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 23056-33-9
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.57
MDL Number MFCD00010688
InChI Key HWZUMEVIIGNXGM-UHFFFAOYSA-N
Synonym 2-chloro-5-nitro-4-picoline, 2-chloro-4-methyl-5-nitro-pyridine, 2-chloro-4-methyl-5-nitro pyridine, 2-chloro-5-nitro-4-methylpyridine, pyridine, 2-chloro-4-methyl-5-nitro, pubchem6223, acmc-209g0j, ksc201s3j, tpc-py010
PubChem CID 345364
IUPAC Name 2-chloro-4-methyl-5-nitropyridine
SMILES CC1=CC(Cl)=NC=C1[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 36°C to 40°C
Color Brown to Cream
Boiling Point 91°C (5.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 172
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2-Chloro-5-nitro-4-picoline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.