Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-nitrobenzoic Acid, 99+%

Catalog No. AC154695000
Click to view available options
Quantity:
100 g
500 g

CAS: 2516-96-3 | C7H4ClNO4 | 201.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2516-96-3
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.56
MDL Number MFCD00007294
InChI Key QUEKGYQTRJVEQC-UHFFFAOYSA-N
Synonym 2-chloro-5-nitrobenzoic acid, benzoic acid, 2-chloro-5-nitro, 6-chloro-3-nitrobenzoic acid, 2-chloro-5-nitro-benzoic acid, 2-chloro-5-nitrobenzoicacid, mesalamine impurity m, pubchem13682, acmc-1cb0i, 2-chloro-5-nitrobenzic acid, 5-nitro-2-chlorobenzoic acid
PubChem CID 17287
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 164°C to 168°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Plastic bottle
Linear Formula ClC6H3(NO2)CO2H
Quantity 500 g
Beilstein 09, 403
Solubility Information Solubility in water: insoluble
Formula Weight 201.56
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitrobenzoic acid, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-739-7
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.