Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-nitropyrimidine 98.0+%, TCI America™
SDP

Catalog No. C25071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25071G 1 g
C25075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25071G Supplier TCI America Supplier No. C25071G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10320-42-0
Molecular Formula C4H2ClN3O2
Molecular Weight (g/mol) 159.529
MDL Number MFCD04117995
InChI Key OFCBNMYNAHUDGE-UHFFFAOYSA-N
PubChem CID 82544
IUPAC Name 2-chloro-5-nitropyrimidine
SMILES C1=C(C=NC(=N1)Cl)[N+](=O)[O-]

Specifications

Melting Point 109°C
Color Yellow
Quantity 1 g
Formula Weight 159.53
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-5-nitropyrimidine
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.