Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-picoline, 94%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
18368-64-4
C6H6ClN
127.57
VXLYOURCUVQYLN-UHFFFAOYSA-N
2-chloro-5-picoline,6-chloro-3-picoline,pyridine, 2-chloro-5-methyl,2-chloro-5-methyl-pyridine,2-chloro-5-methyl pyridine,3-picoline, 6-chloro,pubchem1193,acmc-1bvpj,5-methyl-2-chloropyridine
581393
2-chloro-5-methylpyridine
CC1=CN=C(C=C1)Cl
18368-64-4
C6H6ClN
127.57
VXLYOURCUVQYLN-UHFFFAOYSA-N
2-chloro-5-picoline,6-chloro-3-picoline,pyridine, 2-chloro-5-methyl,2-chloro-5-methyl-pyridine,2-chloro-5-methyl pyridine,3-picoline, 6-chloro,pubchem1193,acmc-1bvpj,5-methyl-2-chloropyridine
581393
2-chloro-5-methylpyridine
CC1=CN=C(C=C1)Cl
Refractive Index 1.5260 to 1.53
Density 1.1690g/mL
Flash Point 90°C
Specific Gravity 1.169
Quantity 5g
Infrared Spectrum Authentic
Color Orange-Pink or Straw-Yellow to Yellow
Packaging Glass bottle
CAS Min % 92.0
CAS Max % 100.0
Assay Percent Range 92% min. (GC)
Formula Weight 127.57
Physical Form Liquid
Percent Purity 94%
Chemical Name or Material 2-Chloro-5-picoline
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning