Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-(trifluoromethyl)pyrazine, 97%

Catalog No. AAH33863MD
Click to view available options
Quantity:
250 mg
1 g

CAS: 799557-87-2 | C5H2ClF3N2 | 182.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 799557-87-2
Molecular Formula C5H2ClF3N2
Molecular Weight (g/mol) 182.53
MDL Number MFCD13189478
InChI Key AIEGIFIEQXZBCP-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl pyrazine, 5-chloro-2-trifluoromethylpyrazine, pyrazine, 2-chloro-5-trifluoromethyl, 2-chloro-5-trifluoromethyl-pyrazine
PubChem CID 18987645
IUPAC Name 2-chloro-5-(trifluoromethyl)pyrazine
SMILES C1=C(N=CC(=N1)Cl)C(F)(F)F

Specifications

Boiling Point 85°C (112 mmHg)
Quantity 250 mg
UN Number UN3267
Formula Weight 182.53
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)pyrazine
Hazard Category H301+H311+H331-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.