Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-(trifluoromethyl)pyridine, 97%

Catalog No. AAL0820722
Click to view available options
Quantity:
100 g

CAS: 52334-81-3 | C6H3ClF3N | 181.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52334-81-3
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
MDL Number MFCD00042225
InChI Key JFZJMSDDOOAOIV-UHFFFAOYSA-N
Synonym 2-chloro-5-trifluoromethyl pyridine, 2,5-ctf, pyridine, 2-chloro-5-trifluoromethyl, ccris 6430, 5-trifluoromethyl-2-chloropyridine, unii-c9g66d966b, 2-chloro-5-trifluoropyridine, 2-chloro-5 trifluoromethyl pyridine
PubChem CID 92342
IUPAC Name 2-chloro-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC=C(Cl)N=C1

Specifications

Melting Point 29°C to 33°C
Density 1.417
Boiling Point 152°C to 153°C
Flash Point 62°C (143°F)
Quantity 100 g
UN Number UN2811
Beilstein 3649688
Formula Weight 181.55
Percent Purity 97%
Chemical Name or Material 2-Chloro-5-(trifluoromethyl)pyridine
Hazard Category H302+H312+H332-H315-H319-H335-H340
Hazard Statement GHS H Statement
H331-H341-H302-H312-H315-H319-H335
Toxic if inhaled.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 257-856-5
RTECSNumber US7530000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.