Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-6-fluoroaniline, 99%

Catalog No. AC355820050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 363-51-9 | C6H5ClFN | 145.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-51-9
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00040309
InChI Key ZJLAWMDJTMMTQB-UHFFFAOYSA-N
Synonym benzenamine, 2-chloro-6-fluoro, 2-chloro-6-fluoro-aniline, 2-chloro-6-fluoro-phenylamine, 6-chloro-2-fluorophenylamine, pubchem1528, 2-fluoro-6-chloroaniline, acmc-209im1, 2-chloro-6-fluoro aniline, 2-chloro-6-fluorobenzenamine, 2-chloro-6-fluorophenylamine
PubChem CID 2734205
IUPAC Name 2-chloro-6-fluoroaniline
SMILES NC1=C(F)C=CC=C1Cl

Specifications

Color Yellow
CAS Min % 98.5
CAS Max % 100.0
Density 1.3160g/mL
Boiling Point 67°C to 69°C (14.0 mmHg)
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5480 to 1.554
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.316
Formula Weight 145.56
Percent Purity 99%
Chemical Name or Material 2-chloro-6-fluoroaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-657-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.