Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-methoxypyridine 98.0+%, TCI America™
SDP

Catalog No. C019725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C019725G 25 g
C01975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C019725G Supplier TCI America Supplier No. C019725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17228-64-7
Molecular Formula C6H6ClNO
Molecular Weight (g/mol) 143.57
MDL Number MFCD00006265
InChI Key VAVGOGHLNAJECD-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-6-methoxy, 2-methoxy-6-chloropyridine, 6-chloro-2-methoxypyridine, 2-chloro-6-methoxy-pyridine, 2-chlor-6-methoxypyridin, pubchem13194, acmc-1cffb, 6-methoxy-2-chloropyridine, 2-chloro-6-methoxy pyridine, 2-chloro-6-methyloxy pyridine
PubChem CID 87009
IUPAC Name 2-chloro-6-methoxypyridine
SMILES COC1=NC(=CC=C1)Cl

Specifications

Color Colorless
Boiling Point 184°C
Quantity 25 g
Formula Weight 143.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-6-methoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.