Learn More
2-Chloro-6-methoxyquinoline-3-carboxaldehyde, 99%
Description
Mainly used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings. As raw material in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Mainly used as solvents. They are used in intermediate steps for the production of paints, plastics, synthetic resins, and dyes. They are also used in the manufacture of perfumes, solvents, and flavorings. As raw material in organic synthesis.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Specifications
Specifications
Melting Point | 146°C to 151°C |
Flash Point | −9°C (16°F) |
Quantity | 25 g |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 221.64 |
Percent Purity | 99% |
Chemical Name or Material | 2-Chloro-6-methoxyquinoline-3-carboxaldehyde |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.