Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-L-mandelic Acid 98.0+%, TCI America™
SDP

Catalog No. C273825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C273825G 25 g
C27385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C273825G Supplier TCI America Supplier No. C273825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52950-19-3
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00798437
InChI Key RWOLDZZTBNYTMS-ZETCQYMHSA-N
Synonym s-+-2-chloromandelic acid, s-2-2-chlorophenyl-2-hydroxyacetic acid, s-2-chloromandelic acid, s-o-chloromandelicacid, 2-chloro-l-mandelic acid, 2s-2-2-chlorophenyl-2-hydroxy-acetic acid, s-2-chlorophenyl hydroxy acetic acid, pubchem5743, s-o-chloromandelic acid, ksc914s2j
PubChem CID 6922874
IUPAC Name (2S)-2-(2-chlorophenyl)-2-hydroxyacetic acid
SMILES C1=CC=C(C(=C1)C(C(=O)O)O)Cl

Specifications

Melting Point 121°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 186.59
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2-Chloro-L-mandelic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.