Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Chloro-L-phenylalanine, 98+%
Click to view available options
Quantity:
250 mg
1 g
Description
The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 196°C to 199°C |
| Solubility Information | Insoluble in water. |
| Quantity | 1 g |
| Formula Weight | 199.64 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 2-Chloro-L-phenylalanine |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction