Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-m-xylene, 98%

Catalog No. AAA1786722
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 6781-98-2 | C8H9Cl | 140.61 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6781-98-2
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000565
InChI Key VDXLAYAQGYCQEO-UHFFFAOYSA-N
Synonym 2-chloro-m-xylene, 2,6-dimethylchlorobenzene, m-xylene, 2-chloro, benzene, 2-chloro-1,3-dimethyl, 2,6-dimethyl chlorobenzene, 2,6-dimethylphenyl chloride, 1-chloro-2,6-dimethylbenzene, 2,6-dimethylchlorobenzene;2-chloro-m-xylene, 2-chloro-meta-xylene, pubchem3643
PubChem CID 32885
IUPAC Name 2-chloro-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)Cl

Specifications

Melting Point -36°C
Density 1.061
Boiling Point 182°C to 183°C
Quantity 100 g
Refractive Index 1.527
Beilstein 2040954
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 140.61
Percent Purity 98%
Chemical Name or Material 2-Chloro-m-xylene
Hazard Category H227-H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 229-843-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.