Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-N-(4-ethoxyphenyl)-4-nitrobenzamide, 97%, Thermo Scientific™

Catalog No. AAH5919003
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328258-90-8
Molecular Formula C15H13ClN2O4
Molecular Weight (g/mol) 320.729
MDL Number MFCD01188651
InChI Key WKNXXIWQPIKWEA-UHFFFAOYSA-N
Synonym 2-chloro-n-4-ethoxyphenyl-4-nitrobenzamide, 2-chloro-n-4-ethoxy-phenyl-4-nitro-benzamide, 2-chloro-4-nitrophenyl-n-4-ethoxyphenyl carboxamide
PubChem CID 711359
IUPAC Name 2-chloro-N-(4-ethoxyphenyl)-4-nitrobenzamide
SMILES CCOC1=CC=C(C=C1)NC(=O)C2=C(C=C(C=C2)[N+](=O)[O-])Cl

Specifications

Quantity 1 g
Formula Weight 320.73
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-(4-ethoxyphenyl)-4-nitrobenzamide
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.