Learn More
2-Chloro-N-(hydroxymethyl)acetamide, 98%
Description
An organic building block. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. As dyestuff intermediate .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
An organic building block. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. As dyestuff intermediate .
Solubility
Very soluble in water. Poorly soluble in aliphatic hydrocarbons.
Notes
Hygroscopic. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
Specifications
Specifications
| Melting Point | 97°C to 103°C |
| Quantity | 10 g |
| UN Number | UN3261 |
| Beilstein | 1811765 |
| Sensitivity | Hygroscopic |
| Solubility Information | Very soluble in water. Poorly soluble in aliphatic hydrocarbons. |
| Formula Weight | 123.54 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Chloro-N- (hydroxymethyl)acetamide |
Safety and Handling
| Hazard Category | H302+H312+H332-H314-H317-H351 |
| Hazard Statement | GHS H Statement H314-H318-H317 Causes severe skin burns and eye damage. Causes serious eye damage. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 220-598-9 |
| RTECSNumber | AB5733000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.