Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-N-(hydroxymethyl)acetamide, 98%

Catalog No. p-7081992
Click to view available options
Quantity:
10 g
50 g

CAS: 2832-19-1 | C3H6ClNO2 | 123.54 g/mol

An organic building block. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. As dyestuff intermediate .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An organic building block. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. As dyestuff intermediate .

Solubility
Very soluble in water. Poorly soluble in aliphatic hydrocarbons.

Notes
Hygroscopic. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2832-19-1
Molecular Formula C3H6ClNO2
Molecular Weight (g/mol) 123.54
MDL Number MFCD00021961
InChI Key TXNSZCSYBXHETP-UHFFFAOYSA-N
Synonym 2-chloro-n-hydroxymethyl acetamide, n-hydroxymethyl-2-chloroacetamide, acetamide, 2-chloro-n-hydroxymethyl, n-methylolchloroacetamide, grota, chloracetamide-n-metholol, n-methylol-chloracetamide, grotan df-35, caswell no. 192bb, chloracetamide-n-methanol
PubChem CID 62466
IUPAC Name 2-chloro-N-(hydroxymethyl)acetamide
SMILES OCNC(=O)CCl

Specifications

Melting Point 97°C to 103°C
Quantity 10 g
UN Number UN3261
Beilstein 1811765
Sensitivity Hygroscopic
Solubility Information Very soluble in water. Poorly soluble in aliphatic hydrocarbons.
Formula Weight 123.54
Percent Purity 98%
Chemical Name or Material 2-Chloro-N- (hydroxymethyl)acetamide
Hazard Category H302+H312+H332-H314-H317-H351
Hazard Statement GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 220-598-9
RTECSNumber AB5733000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.