Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-N-phenylbenzamide, 97%, Thermo Scientific Chemicals

Catalog No. AAH5962303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5962303 1 g
AAH59623MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5962303 Supplier Thermo Scientific Chemicals Supplier No. H5962303

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6833-13-2
Molecular Formula C13H10ClNO
Molecular Weight (g/mol) 231.68
MDL Number MFCD00016336
InChI Key AXQVIWHAEYLGLO-UHFFFAOYSA-N
Synonym 2-chlorobenzanilide, o-chlorobenzanilide, 2-chlorobenzoanilide, benzamide, 2-chloro-n-phenyl, 2-chlorobenzoic acid anilide, benzanilide, 2-chloro, n-phenyl-2-chlorobenzamide, 2-chloro-n-phenyl benzamide, 2-chloro-n-phenyl-benzamide, 2-chlorophenyl-n-benzamide
PubChem CID 23265
IUPAC Name 2-chloro-N-phenylbenzamide
SMILES ClC1=CC=CC=C1C(=O)NC1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 231.68
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Chloro-N-phenylbenzamide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.