Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ 2-Chloro-N6-cyclopentyladenosine
GREENER_CHOICESDP

Catalog No. 170550 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-055-0 50 mg
17-051-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-055-0 Supplier R&D Systems™ Supplier No. 1705/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective A1 agonist

Potent and selective adenosine A1 receptor agonist (Ki values are 0.8, 2300 and 42 nM for human A1, A2A and A3 receptors respectively; EC50 = 18800 nM for hA2B). Centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 37739-05-2
Molecular Formula C15H20ClN5O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 369.81
Purity >98%
Molecular Weight (g/mol) 369.81
Chemical Name or Material 2-Chloro-N6-cyclopentyladenosine
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.