Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-p-cresol 97.0+%, TCI America™
SDP

Catalog No. C200525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C200525G 25 g
C20055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C200525G Supplier TCI America Supplier No. C200525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6640-27-3
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
InChI Key AQJFATAFTQCRGC-UHFFFAOYSA-N
Synonym 2-Chloro-4-methylphenol
PubChem CID 14851
ChEBI CHEBI:79732
IUPAC Name 2-chloro-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)Cl

Specifications

Color Yellow
Boiling Point 196°C
Quantity 25 g
UN Number 2669
Formula Weight 142.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-p-cresol
EINECSNumber (3)-0900
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.