Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzamide, 98%

Catalog No. AAA1570622
Click to view available options
Quantity:
25 g
100 g

CAS: 609-66-5 | C7H6ClNO | 155.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-66-5
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD00007973
InChI Key RBGDLYUEXLWQBZ-UHFFFAOYSA-N
Synonym o-chlorobenzamide, benzamide, 2-chloro, benzamide, o-chloro, o-chloro-benzamide, unii-ke1m5i9i32, chlorobenzamide, pubchem3617, acmc-1aupu, 2-chlorobenzamide, dsstox_cid_30850
PubChem CID 69111
IUPAC Name 2-chlorobenzamide
SMILES NC(=O)C1=CC=CC=C1Cl

Specifications

Melting Point 140°C to 144°C
Quantity 100 g
Beilstein 508510
Formula Weight 155.58
Percent Purity 98%
Chemical Name or Material 2-Chlorobenzamide
Hazard Category H302
Hazard Statement GHS H Statement
H330-H302-H315-H319-H335
Fatal if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 210-195-6
RTECSNumber CV2443737
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.