Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzenesulfonyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2905-23-9
C6H4Cl2O2S
211.07
MFCD00051520
KMVZDSQHLDGKGV-UHFFFAOYSA-N
2-chlorobenzenesulphonyl chloride,2-chlorobenzene-1-sulfonyl chloride,2-chlorobenzenesulfonylchloride,benzenesulfonyl chloride, 2-chloro,o-chlorobenzenesulfonyl chloride,o-chlorobenzenesulfochloride,2-chlorophenylsulfonyl chloride,2-chloro-benzenesulfonyl chloride,benzenesulfonyl chloride, chloro,chloro 2-chlorophenyl sulfone
76187
2-chlorobenzenesulfonyl chloride
C1=CC=C(C(=C1)S(=O)(=O)Cl)Cl
2905-23-9
C6H4Cl2O2S
211.07
MFCD00051520
KMVZDSQHLDGKGV-UHFFFAOYSA-N
2-chlorobenzenesulphonyl chloride,2-chlorobenzene-1-sulfonyl chloride,2-chlorobenzenesulfonylchloride,benzenesulfonyl chloride, 2-chloro,o-chlorobenzenesulfonyl chloride,o-chlorobenzenesulfochloride,2-chlorophenylsulfonyl chloride,2-chloro-benzenesulfonyl chloride,benzenesulfonyl chloride, chloro,chloro 2-chlorophenyl sulfone
76187
2-chlorobenzenesulfonyl chloride
C1=CC=C(C(=C1)S(=O)(=O)Cl)Cl
Specific Gravity 1.548
Refractive Index 1.5745 to 1.5765
Color Orange-Red to Yellow
Quantity 1g
Density 1.5480g/mL
Boiling Point 150.0°C to 152.0°C (12.0mmHg)
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 60°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula ClC6H4SO2Cl
Formula Weight 211.07
Physical Form Liquid or Low Melting Solid
Percent Purity 98%
Chemical Name or Material 2-Chlorobenzenesulfonyl chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

EINECSNumber : 220-807-3