Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorobenzimidazole, 97%

Catalog No. AAA1219306
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAA1219306

Supplier: Thermo Scientific Chemicals A1219306

Only null left
Add to Cart

CAS: 4857-06-1 | C7H5ClN2 | 152.58 g/mol

2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents, alkali metals, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4857-06-1
MDL Number MFCD00051944

Specifications

Melting Point ∼180°C to 185°C
Molecular Formula C16H33Cl
Quantity 5 g
Beilstein 116526
Synonym cetyl chloride, hexadecyl chloride, hexadecane, 1-chloro, n-hexadecyl chloride, chlorohexadecane, unii-wq5zks3ek3, wq5zks3ek3, 1-chloro-hexadecane, 1-chloranylhexadecane, dsstox_cid_7580
Solubility Information Insoluble in water.
InChI Key CLWAXFZCVYJLLM-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCl
IUPAC Name 1-chlorohexadecane
Molecular Weight (g/mol) 260.89
PubChem CID 20993
Formula Weight 152.58
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzimidazole
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-453-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.